Learn More
Medchemexpress LLC HY-106673 10mg Medchemexpress, Hydrocortisone buteprate CAS:72590-77-3 Purity:>98%
SDP
Catalog No. 501932274
Encompass_Preferred
Medchemexpress, HY-106673 10mg Hydrocortisone buteprate CAS:72590-77-3 Hydrocortisone buteprate (Hydrocortisone probutate) is a medium potent, non-halogenated double-ester of hydrocortisone with a favorable benefit/risk ratio for the treatment of inflammatory skin disorders. Hydrocortisone buteprate (Hydrocortisone probutate) is available as a 0.1% cream or ointment formulation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2274 Supplier Medchemexpress LLC Supplier No. HY10667310MG
Medchemexpress, HY-106673 10mg Hydrocortisone buteprate CAS:72590-77-3 Hydrocortisone buteprate (Hydrocortisone probutate) is a medium potent, non-halogenated double-ester of hydrocortisone with a favorable benefit/risk ratio for the treatment of inflammatory skin disorders. Hydrocortisone buteprate (Hydrocortisone probutate) is available as a 0.1% cream or ointment formulation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.