Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone buteprate | 72590-77-3 | 99.3% | 488.6 g/mol | C28H40O7 | 5 MG
SDP

Catalog No. 5000240749
Encompass_Preferred

Hydrocortisone buteprate is a medium-potency, non-halogenated double-ester corticosteroid supplied as a solid analytical standard for laboratory research and analytical applications. The material is provided in a 5 mg package with high purity and is intended for research use only.

  • Solid analytical standard supplied as a 5 mg package.
  • High purity (99.3%).
  • CAS number 72590-77-3.
  • Chemical formula C28H40O7; molecular weight 488.6 g/mol.
  • Used for corticosteroid research and topical formulation studies.
  • Not for human or veterinary use; for laboratory research only.

Catalog No. 50-002-40749 Supplier Medchemexpress LLC Supplier No. HY1066735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrocortisone buteprate is a medium-potency, non-halogenated double-ester corticosteroid supplied as a solid analytical standard for laboratory research and analytical applications. The material is provided in a 5 mg package with high purity and is intended for research use only.

  • Solid analytical standard supplied as a 5 mg package.
  • High purity (99.3%).
  • CAS number 72590-77-3.
  • Chemical formula C28H40O7; molecular weight 488.6 g/mol.
  • Used for corticosteroid research and topical formulation studies.
  • Not for human or veterinary use; for laboratory research only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.