Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone buteprate | 72590-77-3 | 99.3% | 488.61 g/mol | C28H40O7 | 1 ML
SDP

Catalog No. 5000240748
Encompass_Preferred

Hydrocortisone buteprate is a corticosteroid derivative for research use, provided in both solid form and as a DMSO solution for in vitro and in vivo studies. Documented physicochemical and solubility data support formulation and dosing workflows, and the product is intended solely for laboratory research.

  • High reported purity of 99.29%.
  • Molecular formula C28H40O7 and molecular weight 488.61 g/mol.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • Solubility data and in vivo formulation protocols are provided.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-40748 Supplier Medchemexpress LLC Supplier No. HY10667310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hydrocortisone buteprate is a corticosteroid derivative for research use, provided in both solid form and as a DMSO solution for in vitro and in vivo studies. Documented physicochemical and solubility data support formulation and dosing workflows, and the product is intended solely for laboratory research.

  • High reported purity of 99.29%.
  • Molecular formula C28H40O7 and molecular weight 488.61 g/mol.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • Solubility data and in vivo formulation protocols are provided.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.