Learn More
Med Vet International Hydrocortisone Topical Steroid 2.5%, 28gm Tube
Catalog No. 502828788
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Hydrocortisone is a topical steroid that reduces the actions of chemicals in the body that cause inflammation. Topical corticosteroids are indicated for the relief of the inflammatory and pruritic manifestations of corticosteroid-responsive dermatoses. Hydrocortisone topical is used to treat inflammation of the skin caused by a number of conditions such as allergic reactions, eczema, or psoriasis. Hydrocortisone topical will not treat skin infections.
Catalog No. 50-282-8788 Supplier Med Vet International Supplier No. RX45802000403
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Hydrocortisone is a topical steroid that reduces the actions of chemicals in the body that cause inflammation. Topical corticosteroids are indicated for the relief of the inflammatory and pruritic manifestations of corticosteroid-responsive dermatoses. Hydrocortisone topical is used to treat inflammation of the skin caused by a number of conditions such as allergic reactions, eczema, or psoriasis. Hydrocortisone topical will not treat skin infections.