Learn More
Medchemexpress LLC Hydrocortisone cypionate | 508-99-6 | 486.64 | 10 MG
SDP
Catalog No. 5000448811
Encompass_Preferred
Hydrocortisone cypionate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. This product is a solid, appearing white to off-white in color, and is suitable for various research applications. It offers specific solubility characteristics in DMSO and has defined storage conditions to maintain its stability and efficacy.
  • Synthetic glucocorticoid corticosteroid
  • Corticosteroid ester
  • Appears as a solid, white to off-white
  • Solubility in DMSO is 125 mg/mL
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-48811 Supplier Medchemexpress LLC Supplier No. HYU0008910MG
Hydrocortisone cypionate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. This product is a solid, appearing white to off-white in color, and is suitable for various research applications. It offers specific solubility characteristics in DMSO and has defined storage conditions to maintain its stability and efficacy.
  • Synthetic glucocorticoid corticosteroid
  • Corticosteroid ester
  • Appears as a solid, white to off-white
  • Solubility in DMSO is 125 mg/mL
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year