Learn More
Medchemexpress LLC Hydrocortisone cypionate | 508-99-6 | 486.64 | 5 MG
SDP
Catalog No. 5000446578
Encompass_Preferred
Hydrocortisone cypionate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It is intended for research use only.
  • Down-regulates GR transcript in hCMEC/D3 cells in a dose-dependent manner.
  • Supplementation leads to a significant increase in TER across the hCMEC/D3 monolayer.
  • Treated dendritic cells (DCs) exhibit decreased expression of MHC II molecules, costimulatory molecule CD86, and DC-specific marker CD83.
  • Treated dendritic cells (DCs) show strongly reduced IL-12 secretion.
  • Treated dendritic cells (DCs) inhibit IFN-γ production but induce increased release of IL-4.
  • Reduces post-ischemic oxidative stress, perfusion pressure, and transudate formation.
  • Inhibits post-ischemic shedding of syndecan-1, heparan sulfate, and hyaluronan.
  • Inhibits histamine release from mast cells.
Catalog No. 50-004-46578 Supplier Medchemexpress LLC Supplier No. HYU000895MG
Hydrocortisone cypionate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It is intended for research use only.
  • Down-regulates GR transcript in hCMEC/D3 cells in a dose-dependent manner.
  • Supplementation leads to a significant increase in TER across the hCMEC/D3 monolayer.
  • Treated dendritic cells (DCs) exhibit decreased expression of MHC II molecules, costimulatory molecule CD86, and DC-specific marker CD83.
  • Treated dendritic cells (DCs) show strongly reduced IL-12 secretion.
  • Treated dendritic cells (DCs) inhibit IFN-γ production but induce increased release of IL-4.
  • Reduces post-ischemic oxidative stress, perfusion pressure, and transudate formation.
  • Inhibits post-ischemic shedding of syndecan-1, heparan sulfate, and hyaluronan.
  • Inhibits histamine release from mast cells.