Learn More
Medchemexpress LLC Hydrocortisone-d4 | 73565-87-4 | 366.48 | 500 UG
SDP
Catalog No. 5000389053
Encompass_Preferred
Hydrocortisone-d4 is the deuterium-labeled Hydrocortisone. Hydrocortisone (Cortisol) is a steroid hormone or glucocorticoid secreted by the adrenal cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89053 Supplier Medchemexpress LLC Supplier No. HYN0583S2500UG
Hydrocortisone-d4 is the deuterium-labeled Hydrocortisone. Hydrocortisone (Cortisol) is a steroid hormone or glucocorticoid secreted by the adrenal cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.