Learn More
Selleck Chemical LLC Hydrocortisone hemisuccinate
SDP
Catalog No. 5000004196
Encompass_Preferred
Hydrocortisone hemisuccinate (A-hydrocort Hydrocortisone 21-hemisuccinate) a physiological glucocorticoid with anti-inflammatory properties is an inhibitor of proinflammatory cytokine with IC50 of 6 7 M and 21 4 M for Interleukin-6 (IL-6) and IL-3 respectively
Catalog No. 50-000-04196 Supplier Selleck Chemical LLC Supplier No. S597225MG
Add to Cart
Add to Cart
Hydrocortisone hemisuccinate (A-hydrocort Hydrocortisone 21-hemisuccinate) a physiological glucocorticoid with anti-inflammatory properties is an inhibitor of proinflammatory cytokine with IC50 of 6 7 M and 21 4 M for Interleukin-6 (IL-6) and IL-3 respectively