Learn More
Medchemexpress LLC Hydrocortisone hemisuccinate hydrate | 83784-20-7 | 96.42% | 25 MG
SDP
Catalog No. 5000414166
Encompass_Preferred
Hydrocortisone hemisuccinate hydrate is a glucocorticoid and an orally active steroidal anti-inflammatory agent (SAID) with anti-inflammatory and immunomodulatory activities. It inhibits the bioactivity of IL-6 and IL-3 with IC50 values of 6.7 and 21.4 μM, respectively. This compound is suitable for use in the study of ulcerative colitis (UC) and recurrent oral ulcers.
  • Glucocorticoid
  • Orally active steroidal anti-inflammatory agent
  • Inhibits IL-6 and IL-3 bioactivity
  • Used in ulcerative colitis research
  • Used in recurrent oral ulcers research
Catalog No. 50-004-14166 Supplier Medchemexpress LLC Supplier No. 25MGHYB1402A
Hydrocortisone hemisuccinate hydrate is a glucocorticoid and an orally active steroidal anti-inflammatory agent (SAID) with anti-inflammatory and immunomodulatory activities. It inhibits the bioactivity of IL-6 and IL-3 with IC50 values of 6.7 and 21.4 μM, respectively. This compound is suitable for use in the study of ulcerative colitis (UC) and recurrent oral ulcers.
  • Glucocorticoid
  • Orally active steroidal anti-inflammatory agent
  • Inhibits IL-6 and IL-3 bioactivity
  • Used in ulcerative colitis research
  • Used in recurrent oral ulcers research