Learn More
Medchemexpress LLC Hydrocortisone hemisuccinate sodium | 125-04-2 | 99.7% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1402B
Encompass_Preferred
Hydrocortisone hemisuccinate sodium is an orally active physiological glucocorticoid. It inhibits proinflammatory cytokine activity, with IC50s of 6.7 and 21.4 μM for IL-6 and IL-3, respectively. It can be used for the research of ulcerative colitis (UC).
  • Orally active physiological glucocorticoid
  • Inhibits proinflammatory cytokine activity
  • Potential for research in ulcerative colitis (UC)
Catalog No. 50-004-13888