Learn More
Medchemexpress LLC Hydrocortisone hemisuccinate (sodium) | 125-04-2 | 99.7% | 484.51 g/mol | C25H33NaO8 | 10 MG
SDP
Catalog No. 5000297264
Encompass_Preferred
Hydrocortisone hemisuccinate sodium is the sodium salt of hydrocortisone hemisuccinate, a steroidal glucocorticoid used as a research reagent. It is water-soluble and commonly used in anti-inflammatory and glucocorticoid receptor studies, including models of ulcerative colitis. For research use only; not for human consumption.
  • High purity (99.66%) suitable for analytical and biological research.
  • Water-soluble sodium salt for improved aqueous solubility.
  • Reported molecular weight 484.51 g/mol and CAS 125-04-2.
  • Available in small laboratory pack sizes, including 10 mg and solution formats.
  • Datasheet and safety data sheet available for handling and assay information.
Catalog No. 50-002-97264 Supplier Medchemexpress LLC Supplier No. HYB1402B10MG
Hydrocortisone hemisuccinate sodium is the sodium salt of hydrocortisone hemisuccinate, a steroidal glucocorticoid used as a research reagent. It is water-soluble and commonly used in anti-inflammatory and glucocorticoid receptor studies, including models of ulcerative colitis. For research use only; not for human consumption.
  • High purity (99.66%) suitable for analytical and biological research.
  • Water-soluble sodium salt for improved aqueous solubility.
  • Reported molecular weight 484.51 g/mol and CAS 125-04-2.
  • Available in small laboratory pack sizes, including 10 mg and solution formats.
  • Datasheet and safety data sheet available for handling and assay information.