Learn More
Medchemexpress LLC Hydrocortisone phosphate sodium | 6000-74-4 | MFCD00152562 | 99.1% | 486.40 g·mol⁻¹ | C21H29Na2O8P | 50 MG
SDP
Catalog No. 5000241564
Encompass_Preferred
Hydrocortisone phosphate sodium is the disodium phosphate salt of hydrocortisone, a water-soluble corticosteroid used as a research reagent. It has corticosteroid activity with relatively low anti-inflammatory potency and a short duration of action, and is used in biochemical and pharmacological studies, including research on ocular conditions and gastric ulcers.
  • Water-soluble sodium phosphate salt suitable for aqueous formulations.
  • Useful for biochemical and pharmacological research of corticosteroid activity.
  • Relatively low anti-inflammatory potency with short duration of action.
  • High purity and defined molecular weight for reproducible results.
  • Stable when stored sealed and away from moisture; follow cold storage recommendations for solutions.
Catalog No. 50-002-41564 Supplier Medchemexpress LLC Supplier No. HY10565650MG
Hydrocortisone phosphate sodium is the disodium phosphate salt of hydrocortisone, a water-soluble corticosteroid used as a research reagent. It has corticosteroid activity with relatively low anti-inflammatory potency and a short duration of action, and is used in biochemical and pharmacological studies, including research on ocular conditions and gastric ulcers.
  • Water-soluble sodium phosphate salt suitable for aqueous formulations.
  • Useful for biochemical and pharmacological research of corticosteroid activity.
  • Relatively low anti-inflammatory potency with short duration of action.
  • High purity and defined molecular weight for reproducible results.
  • Stable when stored sealed and away from moisture; follow cold storage recommendations for solutions.