Learn More
Medchemexpress LLC Hydrocortisone sodium phosphate | 6000-74-4 | MFCD00152562 | 99.1% | 486.40 g·mol⁻¹ | C21H29Na2O8P | 100 MG
SDP
Catalog No. 5000241628
Encompass_Preferred
Hydrocortisone sodium phosphate is the sodium phosphate ester of hydrocortisone, a short-acting glucocorticoid corticosteroid supplied as a white to off-white solid. Its water-soluble form is suitable for biochemical, pharmacological, and formulation studies where a soluble corticosteroid derivative is required.
  • High purity: 99.1%.
  • Water soluble for easy formulation and in vitro use.
  • Molecular weight 486.40 g·mol⁻¹.
  • White to off-white solid appearance.
  • Provided in small pack sizes suitable for laboratory research.
Catalog No. 50-002-41628 Supplier Medchemexpress LLC Supplier No. HY105656100MG
Hydrocortisone sodium phosphate is the sodium phosphate ester of hydrocortisone, a short-acting glucocorticoid corticosteroid supplied as a white to off-white solid. Its water-soluble form is suitable for biochemical, pharmacological, and formulation studies where a soluble corticosteroid derivative is required.
  • High purity: 99.1%.
  • Water soluble for easy formulation and in vitro use.
  • Molecular weight 486.40 g·mol⁻¹.
  • White to off-white solid appearance.
  • Provided in small pack sizes suitable for laboratory research.