Learn More
Medchemexpress LLC Hydrocortisone sodium phosphate | 6000-74-4 | MFCD00152562 | 99.1% | 486.40 g/mol | C21H29Na2O8P | 25 MG
SDP
Catalog No. 5000241056
Encompass_Preferred
Hydrocortisone sodium phosphate is the sodium phosphate ester of hydrocortisone, a water-soluble corticosteroid with glucocorticoid anti-inflammatory activity and a short duration of action. It is supplied as a solid research reagent for biochemical and pharmacological studies; consult the safety data sheet for handling and storage.
  • Water-soluble sodium phosphate ester of hydrocortisone.
  • Provides glucocorticoid anti-inflammatory activity with short duration of action.
  • High reported purity supports analytical and biological experiments.
  • Suitable for in vitro and in vivo research applications.
  • Packaged as a 25 mg solid for convenient small-scale use.
Catalog No. 50-002-41056 Supplier Medchemexpress LLC Supplier No. HY10565625MG
Hydrocortisone sodium phosphate is the sodium phosphate ester of hydrocortisone, a water-soluble corticosteroid with glucocorticoid anti-inflammatory activity and a short duration of action. It is supplied as a solid research reagent for biochemical and pharmacological studies; consult the safety data sheet for handling and storage.
  • Water-soluble sodium phosphate ester of hydrocortisone.
  • Provides glucocorticoid anti-inflammatory activity with short duration of action.
  • High reported purity supports analytical and biological experiments.
  • Suitable for in vitro and in vivo research applications.
  • Packaged as a 25 mg solid for convenient small-scale use.