Learn More
Medchemexpress LLC Hydrocortisone (Standard) | 50-23-7 | 362.46 | 100 MG
SDP
Catalog No. 5000388913
Encompass_Preferred
Hydrocortisone (Standard) is the analytical standard of Hydrocortisone. This product is intended for research and analytical applications. Hydrocortisone (Cortisol) is a steroid hormone or glucocorticoid secreted by the adrenal cortex.
  • Analytical standard
  • Intended for research and analytical applications
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
  • Steroid hormone or glucocorticoid secreted by the adrenal cortex
Catalog No. 50-003-88913 Supplier Medchemexpress LLC Supplier No. HYN0583R100MG
Hydrocortisone (Standard) is the analytical standard of Hydrocortisone. This product is intended for research and analytical applications. Hydrocortisone (Cortisol) is a steroid hormone or glucocorticoid secreted by the adrenal cortex.
  • Analytical standard
  • Intended for research and analytical applications
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
  • Steroid hormone or glucocorticoid secreted by the adrenal cortex