Learn More
STEMCELL Technologies Hydrocortisone Stock Solution, Size: 30 mL
SDP
Catalog No. NC1062339
Encompass
Hydrocortisone Stock Solution is recommended for use as a cell culture supplement and for the preparation of PneumaCult™, MammoCult™, and EpiCult™ media when culturing airway epithelial cells, mammary primary cells, or tumor cell lines.Hydrocortisone Stock Solution is provided at a concentration of 96 µg/mL, which is ideal as a 200X stock solution for use with PneumaCult™-ALI (Catalog #05001), MammoCult™ (Catalog #05620), EpiCult™-B (Human; Catalog #05601), and EpiCult™-C (Catalog #05630) media. It may also be used for preparing PneumaCult™-Ex Medium (Catalog #05008) or PneumaCult™-Ex Plus Medium (Catalog #05040).
Catalog No. NC1062339 Supplier STEMCELL Technologies Supplier No. 07926
Hydrocortisone Stock Solution is recommended for use as a cell culture supplement and for the preparation of PneumaCult™, MammoCult™, and EpiCult™ media when culturing airway epithelial cells, mammary primary cells, or tumor cell lines.Hydrocortisone Stock Solution is provided at a concentration of 96 µg/mL, which is ideal as a 200X stock solution for use with PneumaCult™-ALI (Catalog #05001), MammoCult™ (Catalog #05620), EpiCult™-B (Human; Catalog #05601), and EpiCult™-C (Catalog #05630) media. It may also be used for preparing PneumaCult™-Ex Medium (Catalog #05008) or PneumaCult™-Ex Plus Medium (Catalog #05040).