Learn More
AdipoGen Hydrocupreine
SDP
Catalog No. 502560681
Encompass_Preferred
Chemical. CAS 5962-19-6. Formula C19H24N2O2. MW 312.41. Hydrocupreine 6-Hydroxydihydrocinchonidine is a derivative of cinchonidine and metabolite of quinine, alkaloids found in the bark of the Cinchona tree. It has been shown to have antimicrobial and antitumor activity, and inhibitory effect on Trypanosoma brucei and Streptococcus pneumoniae. Hydrocupreine can be used as a building block, starting material or reagent in organic synthesis to create more complex chiral chemical compounds and/or azo dyes. It has been used in organocatalaysis and during Friedel-Crafts amination.
Catalog No. 50-256-0681 Supplier AdipoGen Supplier No. CDXH0927G005
Chemical. CAS 5962-19-6. Formula C19H24N2O2. MW 312.41. Hydrocupreine 6-Hydroxydihydrocinchonidine is a derivative of cinchonidine and metabolite of quinine, alkaloids found in the bark of the Cinchona tree. It has been shown to have antimicrobial and antitumor activity, and inhibitory effect on Trypanosoma brucei and Streptococcus pneumoniae. Hydrocupreine can be used as a building block, starting material or reagent in organic synthesis to create more complex chiral chemical compounds and/or azo dyes. It has been used in organocatalaysis and during Friedel-Crafts amination.