Learn More
Vector Laboratories Hydrodabcyl N-succinimidyl ester, Hydrodabcyl-ONSu, 25 mg
SDP
Supplier:  Vector Laboratories EBR003
Encompass
Hydrodabcyl N-succinimidyl ester, Hydrodabcyl-ONSu, is an active ester (activated hydrodabcyl) that readily reacts with primary and secondary amides at room temperature and can be stored for long periods of time.
Catalog No. NC2738082