Learn More
Selleck Chemical LLC Hydroferulic acid S5135-25mg
SDP
Catalog No. 503112317
Encompass_Preferred
Hydroferulic acid (Dihydroferulic acid Dihydroconiferylic acid) is classified as a member of the phenylpropanoic acids It is slightly soluble in water and acidic
Catalog No. 50-311-2317 Supplier Selleck Chemical LLC Supplier No. S513525MG
Add to Cart
Add to Cart
Hydroferulic acid (Dihydroferulic acid Dihydroconiferylic acid) is classified as a member of the phenylpropanoic acids It is slightly soluble in water and acidic