Learn More
Cayman Chemical Hydroflumethiazide-13C-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 1189877-11-9, 1 mg
SDP
Catalog No. 502988049
Encompass_Preferred
An internal standard for the quantification of hydroflumethiazide by GC- or LC-MS
Catalog No. 50-298-8049 Supplier Cayman Chemical Supplier No. 316081 MG
An internal standard for the quantification of hydroflumethiazide by GC- or LC-MS