Learn More
Cayman Chemical HydroflumethIazIde 50mg
SDP
Catalog No. 502994131
Encompass_Preferred
A thiazide diuretic; inhibitor of carbonic anhydrase (CA); selective for CAII, VB, VII, IX, XII, and XIV (Kis = 305-235 nM) over CAI, IV, and VI (Kis = 2,840-8,250 nM) and CAIII, VA, and XIII (Kis = >10 µM)
Catalog No. 50-299-4131 Supplier Cayman Chemical Supplier No. 2361250 MG
A thiazide diuretic; inhibitor of carbonic anhydrase (CA); selective for CAII, VB, VII, IX, XII, and XIV (Kis = 305-235 nM) over CAI, IV, and VI (Kis = 2,840-8,250 nM) and CAIII, VA, and XIII (Kis = >10 µM)