Learn More
LAB ALLEY Lab Alley Hydrofluoric Acid 5% Solution Lab Grade
SDP
Catalog No. NC2828729
Encompass
Hydrofluoric Acid also known as Hydrogen Fluoride Fluoric Acid Fluorine Hydride and Fluor Hydric Acid appears as a clear colorless to yellow liquid. It is a fuming liquid with an acidic strong suffocating odor. It is shipped as a compressed gas in steel cylinders and is soluble in Water and Alcohol but slightly soluble in Ether. It is the strongest inorganic acid and is commonly used in research due to its ability to etch silicon compounds. Lab Alley Hydrofluoric Acid contains 5% Hydrogen Fluoride mixed with 95% of Water. Chemically pure or Laboratory reagents are two terms often used to describe Lab Grade chemicals. Lab Grade chemicals do not meet any accepted quality or purity requirements such as the ACS Grade the USP Grade and the FCC Grade despite their acceptable purity.
Catalog No. NC2828729 Supplier LAB ALLEY Supplier No. C4354500ML
Hydrofluoric Acid also known as Hydrogen Fluoride Fluoric Acid Fluorine Hydride and Fluor Hydric Acid appears as a clear colorless to yellow liquid. It is a fuming liquid with an acidic strong suffocating odor. It is shipped as a compressed gas in steel cylinders and is soluble in Water and Alcohol but slightly soluble in Ether. It is the strongest inorganic acid and is commonly used in research due to its ability to etch silicon compounds. Lab Alley Hydrofluoric Acid contains 5% Hydrogen Fluoride mixed with 95% of Water. Chemically pure or Laboratory reagents are two terms often used to describe Lab Grade chemicals. Lab Grade chemicals do not meet any accepted quality or purity requirements such as the ACS Grade the USP Grade and the FCC Grade despite their acceptable purity.