Learn More
Hydrogen chloride, 1M in diethyl ether
Description
In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, s, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Concentration | 1M in diethyl ether |
| Density | 0.731 |
| Refractive Index | 1.3535 |
| Quantity | 100 mL |
| Flash Point | −40°C (−40°F) |
| UN Number | UN2924 |
| Sensitivity | Air and light sensitive |
| Solubility Information | Fully misicible in water. |
| Formula Weight | 36.46 |
| Chemical Name or Material | Hydrochloric acid |
Safety and Handling
| Hazard Category | H224-H302-H314-H335-H336 |
| Hazard Statement | GHS H Statement H225-H331-H314-H302-H336 Highly flammable liquid and vapor. Toxic if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.