Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride – 2-propanol solution, Solstice

Catalog No. 60048465 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-465 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-465 Supplier Solstice Supplier No. 350091L

0.1 M HCl in 2-propanol (0.1N), for Total Base Number, according to DIN 51558-1, volumetric

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 67-63-0
MDL Number MFCD00011324
PubChem CID 313
ChEBI CHEBI:17883

Specifications

Linear Formula HCl · CH3CH(OH)CH3
UN Number UN2924
Beilstein 1098214
Formula Weight 36.46g/mol
Quantity 1 L
Flash Point 12°C (53.6°F)
Chemical Name or Material Hydrogen chloride - 2-propanol solution
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
DOTInformation 8(3)/PGII
RTECSNumber 200-661-7
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.