Learn More
Hydrogen chloride, 2M in diethyl ether
Description
Hydrogen chloride, 2M in diethyl ether is used in organic synthesis to prepare emodin derivatives and polyamine derivatives of betulinic acid and beta-sitosterol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Concentration | 2M in diethyl ether |
| Density | 0.747 |
| Quantity | 500 mL |
| Flash Point | −40°C (−40°F) |
| UN Number | UN2924 |
| Sensitivity | Air and light sensitive |
| Solubility Information | Miscible with methanol,ethanol and ether. |
| Formula Weight | 36.46 |
| Chemical Name or Material | Hydrochloric acid |
Safety and Handling
| Hazard Category | H224-H302-H314-H335-H336 |
| Hazard Statement | GHS H Statement H225-H331-H314-H302-H336 Highly flammable liquid and vapor. Toxic if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| EINECSNumber | 231-595-7 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.