Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride, 4M in 1,4-dioxane, 99%

Catalog No. p-7023412
Change view
Click to view available options
Quantity:
0.1 mol
0.5 mol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16935JQ 0.1 mol
AAA16935JT 0.5 mol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16935JQ Supplier Thermo Scientific Chemicals Supplier No. A16935JQ
Only null left
Add to Cart

CAS: 7647-01-0 | ClH | 36.46 g/mol

Hydrogen chloride, 4M in 1,4-dioxane is used in the preparation of N-methyl-O-benzoylhydroxylamine hydrochloride via reaction with benzoic acid, carbonyldiimidazole and N-methylhydroxylamine hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Linear Formula HCl
UN Number UN2924
Concentration 4M in 1,4-dioxane
Density 1.05
Solubility Information Miscible with alcohol,ether and 1,4-dioxane.
IUPAC Name chlorane
Formula Weight 36.46
Percent Purity 99%
Quantity 0.1 mol
Flash Point 17°C (63°F)
Sensitivity Hygroscopic
Chemical Name or Material Hydrochloric acid
Show More Show Less
Hazard Category H225-H314-H331-H335-H350
Hazard Statement GHS H Statement
H225-H331-H314-H351-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.