Learn More
Hydrogen chloride, 5 to 6M soln. in 2-propanol, pure
Description
Hydrogen chloride, 5 to 6M soln. in 2-propanol may be used in measuring their conductance at 250. It is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Concentration | 5 to 6M soln. in 2-propanol |
| Density | 0.909 |
| Odor | Alcohol-like |
| Linear Formula | HCl |
| Quantity | 500 mL |
| Flash Point | 11°C (52°F) |
| UN Number | UN2924 |
| Sensitivity | Hygroscopic |
| Solubility Information | Miscible with water,methanol,ethanol and 2-propanol. |
| IUPAC Name | hydrogen chloride |
| Show More |
Safety and Handling
| Hazard Category | H225-H314-H332-H335-H336 |
| Hazard Statement | GHS H Statement H225-H314-H336 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: I |
| EINECSNumber | 231-595-7 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.