Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride, ca 0.5M soln. in methanol

Catalog No. AAH31570AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31570AP 500 mL
AAH31570AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31570AP Supplier Thermo Scientific Chemicals Supplier No. H31570AP
  • $149.50
    $138.65 / Each of 1

    Save $10.85

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

CAS: 7647-01-0 | ClH | 36.46 g/mol

Industrially used in the production of inorganic salts of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, pharmaceutical hydrochloride, Also as catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Manufacture of fertilizers, Hydrolysing of starch and proteins in the preparation of various food products, Refining ore Leather deliming / tanning agents, Oil- and gas-well treatment, Textile scouring agents, Removing scale from boilers and heat-exchange equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Linear Formula HCl
UN Number UN1992
Concentration ≈0.5M soln. in methanol
Density 0.8
Solubility Information Miscible in water823g/L (0°C),673g/L (30°C). HCl dissolves readily into ice. Reacts exothermically with organic bases (amines,amides) and inorganic bases (oxides and hydroxides of metals).
Formula Weight 36.46
Quantity 500 mL
Flash Point 12°C (54°F)
Chemical Name or Material Hydrochloric acid
Hazard Category H225-H301+H311+H331-H315-H319-H335-H336-H370-H372
Hazard Statement GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.