Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrogen chloride solution 1.0 M in acetic acid | 7647-01-0 | MFCD00011324 | 100ML
Catalog No. 501880609
Encompass
Hydrogen chloride solution 1.0 M in acetic acid | Purity: 950-1100 mmol/L (silver nitrate titration) | Mol Wt: 36.46 | 7647-01-0 | MFCD00011324 | 100ML
Catalog No. 50-188-0609 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 304174100ML
Hydrogen chloride solution 1.0 M in acetic acid | Purity: 950-1100 mmol/L (silver nitrate titration) | Mol Wt: 36.46 | 7647-01-0 | MFCD00011324 | 100ML