Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydrogen Chloride Solution

Catalog No. 705551100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-555-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 70-555-1100ML Supplier Sigma Aldrich Supplier No. 705551100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydrogen chloride solution can be used in the Pinner synthesis of imidate hydrochlorides, which involves acid-catalyzed condensation of a nitrile and an alcohol under anhydrous conditions.

Specifications

CAS 67950-05-4
Melting Point 80°C to 85°C
Molecular Formula C18H22ClP
Linear Formula C18H22ClP
MDL Number MFCD15146311
Quantity 100 mL
Synonym Chlorodimesitylphosphine
Molecular Weight (g/mol) 304.79 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.