Learn More
World Precision Instrument Hydrogen Electrode Tip, Replacement
SDP
Catalog No. 502521043
Encompass
Replacement tips
Set of 3 hydrogen replacement tips for ion selective electrodes. Change the tip on your holder in less than a minute.
Catalog No. 50-252-1043 Supplier World Precision Instrument Supplier No. TIPH
Replacement tips
Set of 3 hydrogen replacement tips for ion selective electrodes. Change the tip on your holder in less than a minute.