Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydrogen Fluoride Pyridine

Catalog No. 18422525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-422-525G 25 g
18-422-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 18-422-525G Supplier Sigma Aldrich Supplier No. 18422525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

May contain or for mLow levels of calcium fluoride during storage. Presence of this does not impact the specification values.Reactant for preparation of:Epimers of shikimic acid with the features of fucosylated glycans via zinc-mediated reductive ring opening followed by a Barbier reactionVaccinia H1-related (VHR) phosphatase inhibitor with a nonacidic phosphate-mimicking core structureCandidates for nucleic acid drugs-d-substituted gem-difluoroalkanes by oxidative desulfurization-difluorinationReagent for:Halofluorination reactionsSure/Pac is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 62778-11-4
Density 1.1 g/mL at 20°C
MDL Number MFCD00012436
Quantity 25 g
Synonym HF-Pyridine; Olah's reagent; PPHF; Poly(pyridine fluoride); Pyridine hydrofluoride; Pyridinium poly(hydrogen fluoride); Pyridinium polybifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.