Hydrogen Peroxide 6% (Stabilized) is composed of 6% hydrogen peroxide (H2O2) and 94% water (H2O) with trace amount of acetic acid. Lab Grade Hydrogen Peroxide 6% is a clear colorless liquid that contains stabilizers. These stabilizers help prevent the breakdown of hydrogen peroxide and prolong the chemical composition. Hydrogen peroxide is a great solution for disinfecting applications. Laboratory Grade Hydrogen Peroxide 6% contains intermediate purity and is suitable for educational or research laboratories while it is not safe to use in foods drugs or medicines. Lab Alley has a consistent quality product and are the reliable choice for your Laboratory Grade Hydrogen Peroxide 6% solution needs. With quick processing at Lab Alley is where to buy Lab Grade Hydrogen Peroxide 6%. Lab grade hydrogen peroxide 6% is in stock in multiple sizes ranging from 500mL bottles to 55 gallon drums. We also provide pallets of chemicals.