Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 0.3, ULTREX™ II Ultrapure Reagent, J.T. Baker™

Catalog No. 02003185 Shop All Avantor Products
Change view
Click to view available options
Quantity:
450 mL
Packaging:
HDPE Pressure Bottom Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-185 450 mL HDPE Pressure Bottom Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-185 Supplier Avantor Supplier No. 515501
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Reagents for critical elemental analysis with less than 10 parts-per-trillion (ppt) levels of up to 65 elements

    For use in ICP (Inducutively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Packaging HDPE Pressure Bottom Poly Bottle
Quantity 450 mL
Specific Gravity 1L = 1.11kg
Formula Weight 34.01
Percent Purity 30%
Grade ULTREX™ II Ultrapure Reagent
Chemical Name or Material Hydrogen Peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.