Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 10% (w/w), Reagent Grade, Ricca Chemical
SDP

Catalog No. R38213104BV Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Vented Black Poly Bottle
Vented Opaque Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R38213104BV 4 L Vented Black Poly Bottle
R38213101BV 1 L Vented Opaque Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R38213104BV Supplier Ricca Chemical Company Supplier No. R38213104BV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stabilized with Acetanilide

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Percent Purity 10%
Grade Reagent
Packaging Vented Black Poly Bottle
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 10% (w/w) Aqueous Solution, Stabilized with Acetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.