Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Hydrogen Peroxide Solution, 10% w/w (Size: 1L)
SDP

Catalog No. NC3705953
Encompass_Preferred

Hydrogen Peroxide Solution, 10% w/w (Size: 1L) CAS# 7722-84-1

Catalog No. NC3705953 Supplier Aqua Solutions Supplier No. H30851L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrogen Peroxide Solution, 10% w/w (Size: 1L) CAS# 7722-84-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.