Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Hydrogen Peroxide 5% w/w Solution, Lab Grade (1L)
SDP

Catalog No. NC0239415
Encompass_Preferred

Hydrogen Peroxide 5% w/w Solution, Lab Grade (1L)

Catalog No. NC0239415 Supplier Aqua Solutions Supplier No. H30791L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrogen Peroxide 5% w/w Solution, Lab Grade (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.