Learn More
ECOLAB INC Hydrogen Peroxide Sterilant, 35% w/w concentration, 150 ml bottle, 24 bottles per case, 6-log sporicidal efficacy, RFID traceability, residue-free
Supplier:  ECOLAB INC 6601055
Encompass
Hydrogen Peroxide Sterilant, 35% w/w concentration, 150 ml bottle, 24 bottles per case, 6-log sporicidal efficacy, RFID traceability, residue-free
Catalog No. NC1097261