Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 3% Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S031091 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
3.8 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031091 500 mL
S031136 3.8 L
S031066 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S031091 Supplier Aldon Corporation Supplier No. IS17035
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-84-1

Specifications

Boiling Point Approx. 100°C
Percent Purity 3%
Grade Reagent
Packaging Amber HDPE
pH 4.5
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.