Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Hydrogen Peroxide 3% w/w Reagent Grade (4L)
SDP

Catalog No. NC9706927
Encompass_Preferred

Hydrogen Peroxide 3% w/w Reagent Grade (4L)

Catalog No. NC9706927 Supplier Aqua Solutions Supplier No. H30754L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrogen Peroxide 3% w/w Reagent Grade (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.