Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Hydrogen Peroxide 3% w/w Reagent Grade (4L)
SDP

Supplier:  Aqua Solutions H30754LC

Encompass_Preferred

Hydrogen Peroxide 3% w/w Reagent Grade (4L)

Catalog No. NC9202303


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.