Learn More
Hach Company Hydrogen Peroxide Solution, 30%, 473 mL (No Air Shipments)
Catalog No. 501994176 Shop All Hach Company Products
Encompass
ACS Grade, CAS No. 7722-84-1. H2O2, 30%, FW = 34.01. For Digesdahl digestions
Catalog No. 50-199-4176 Supplier Hach Company Supplier No. 14411
ACS Grade, CAS No. 7722-84-1. H2O2, 30%, FW = 34.01. For Digesdahl digestions