Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™

Catalog No. H325100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
30 gal.
Packaging:
Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H325-100 100 mL Poly Bottle
H325-500 500 mL Poly Bottle
H325-4 4 L Poly Bottle
H325-30GAL 30 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. H325-100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7722-84-1 | Molecular Formula: H2O2 | Molecular Weight: 34.014 g/mol

Hydrogen Peroxide, 30% (Certified ACS)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 108°C
Melting Point -33°C
Density 1.110g/cm³
Identification Pass test
Packaging Poly Bottle
Residue after Evaporation 0.002 % max.
Vapor Pressure 23mmHg at 30°C
Assay Percent Range 29.0 to 32.0 %
Decomposition Information Decomposes information: >125°C
pH 3.3
Quantity 100 mL
Physical Form Liquid
Formula Weight 34
Percent Purity 29.0 to 32.0%
Grade ACS
Chemical Name or Material Hydrogen Peroxide, 30%
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe eye irritation and possible burns. Irritating to skin. Harmful if swallowed.
NFPA
Health:3 Flammability:0 Instability:1
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.