Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30% Solution, AR, For ACS analysis, Macron Fine Chemicals™

Catalog No. 14650383 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
8 pt.
Packaging:
Plastic Bottle
Poly Container
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
14-650-383 500 mL Plastic Bottle
MK-5240-8 8 pt. Poly Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-650-383 Supplier Avantor Supplier No. 524005
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Assay Percent Range 29.0 to 32.0 %
Packaging Plastic Bottle
Quantity 500 mL
Formula Weight 34.01
Grade For ACS analysis
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 30% Solution, ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.