Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30%, Solution, Reagent, 29.0-32.0%, Spectrum™ Chemical
SDP

Catalog No. 18610954 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-954 500 mL Poly Bottle
18-610-952 200 L Drum
18-610-953 100 mL Poly Bottle
18-610-961 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-954 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H1070500MLPL
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Hydrogen Peroxide
CAS 7722-84-1, 7732-18-5
CAS Max % 30, 70
Percent Purity 29 to 32%
Grade Reagent
Packaging Poly Bottle
Residue after Evaporation 0.002%
Specific Gravity 1.1
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.