Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30% (Stabilized with Sodium Stannate/Certified), Fisher Chemical

Catalog No. H323500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H323-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. H323-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5 , 12058-66-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Undesignated
Boiling Point 108°C
Melting Point -33°C
Assay Percent Range 29 to 36 %
UN Number 2014
Formula Weight 34.01
Percent Purity 29 to 36%
Grade Certified
Concentration or Composition (by Analyte or Components) Stabilized with Sodium Stannate
Identification Pass Test
Packaging Poly Bottle
Residue after Evaporation 0.02% max.
Vapor Pressure 23mmHg at 30°C
pH 3.3
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 30%, Stabilized with Sodium Stannate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.