Learn More
Hydrogen Peroxide, 35% (w/w), Reagents
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 152°C |
| Percent Purity | 35% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.1258 |
| Quantity | 4 L |
| Sterility | Non-sterile |
| Physical Form | Liquid |
| Chemical Name or Material | Hydrogen Peroxide, 35% (w/w) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.