Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 35% (w/w), Reagents

Catalog No. 01334973
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-973 1 L
01-334-971 20 L
01-334-970 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-973 Supplier Reagents Holdings Llc Supplier No. CH3020001A
 Restricted Item
Add to Cart
Add to Cart

Hydrogen Peroxide, 35% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Dioxide, Oxydol
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 152°C
Percent Purity 35%
Grade Technical
Packaging Poly Bottle
Specific Gravity 1.1258
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 35% (w/w)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.