Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 4% (w/w), Reagents

Hydrogen Peroxide, 4% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CH2995004A

Quantity 4 L

Catalog No. 01-334-975


Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 7722-84-1
18.02
Dihydrogen Dioxide, Oxydol
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

Colorless
100°C
Dihydrogen Dioxide, Oxydol
O
18.02
Reagent
1.0146
Non-sterile
Hydrogen Peroxide, 4% (w/w)
7732-18-5 , 7722-84-1
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
4%
Poly Bottle
4 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.