Learn More
Santa Cruz Biotechnology Hydrogen Peroxide,7722-84-1,500mL
Catalog No. NC1434807
Encompass
Molecular Formula H2O2, Molecular Weight 34.01, Synonyms oxydol; perhydrol, Hydrogen peroxide (H2O2) is an incredibly ubiquitous oxidant used as a reagent, disinfectant, antiseptic, and propellant. In synthetic chemistry, H2O2 is used for epoxidation and hydroxylation transformations, and is used as the raw oxidant source for generating more complicated and specific organic/inorganic peroxides. H2O2 in aqueous solution is used as a laboratory disinfectant and as a topical antiseptic. The disinfectant, bleaching, and non-toxic properties of H2O2 provide a wide scope of industrial and commercial applications.
Catalog No. NC1434807 Supplier Santa Cruz Biotechnology Supplier No. SC203336A
 Restricted Item
Add to Cart
Add to Cart
Molecular Formula H2O2, Molecular Weight 34.01, Synonyms oxydol; perhydrol, Hydrogen peroxide (H2O2) is an incredibly ubiquitous oxidant used as a reagent, disinfectant, antiseptic, and propellant. In synthetic chemistry, H2O2 is used for epoxidation and hydroxylation transformations, and is used as the raw oxidant source for generating more complicated and specific organic/inorganic peroxides. H2O2 in aqueous solution is used as a laboratory disinfectant and as a topical antiseptic. The disinfectant, bleaching, and non-toxic properties of H2O2 provide a wide scope of industrial and commercial applications.